siRNA Details
Gene Symbol: Rasgef1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029004.1 | NP_083280.1 | 7 | 894 | ||
GenBank mRNA | AK018593.1 | - | 7 | 883 | ||
AK039330.1 | - | 1258 | ||||
AK047088.1 | - | 6 | 773 |
