siRNA Details
| Gene Symbol: Mlkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029005.2 | NP_083281.1 | 5, 6 | 859 | ||
| GenBank mRNA | AK018636.1 | - | 5, 6 | 1135 | ||
| AK170260.1 | - | 5, 6 | 860 | |||
| BC023755.1 | - | 5, 6 | 1144 | 5354568 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlkl| Gene Symbol: Mlkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029005.2 | NP_083281.1 | 5, 6 | 859 | ||
| GenBank mRNA | AK018636.1 | - | 5, 6 | 1135 | ||
| AK170260.1 | - | 5, 6 | 860 | |||
| BC023755.1 | - | 5, 6 | 1144 | 5354568 | ||