siRNA Details
| Gene Symbol: Abca12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175210.3 | NP_780419.2 | 11, 12 | 1615 | ||
| GenBank mRNA | AK028781.1 | - | 11, 12 | 1616 | ||
| AK029031.1 | - | 11, 12 | 1616 | |||
| AK036764.1 | - | 7, 8 | 862 | |||
| BC158053.1 | - | 11, 12 | 1428 | 9088162 | ||
| BC158063.1 | - | 11, 12 | 1428 | 9007362 | ||

Abca12
Genomic Map