siRNA Details
| Gene Symbol: Abca12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175210.3 | NP_780419.2 | 3, 4 | 636 | ||
| GenBank mRNA | AK028781.1 | - | 3, 4 | 637 | ||
| AK029031.1 | - | 3, 4 | 637 | |||
| BC158053.1 | - | 3, 4 | 449 | 9088162 | ||
| BC158063.1 | - | 3, 4 | 449 | 9007362 | ||

Abca12
Genomic Map