siRNA Details
| Gene Symbol: Wdr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159527.1 | NP_001152999.1 | 24 | 3100 | ||
| NM_172470.3 | NP_766058.3 | 25 | 3133 | |||
| GenBank mRNA | AK029896.1 | - | 24 | 3055 | ||
| AK083971.1 | - | 24 | 3045 | |||
| BC056925.1 | - | 25 | 3242 | 5704780 | ||

Wdr35
Genomic Map