siRNA Details
| Gene Symbol: Ccdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029061.3 | NP_083337.1 | 7, 8 | 900 | ||
| GenBank mRNA | AK015811.1 | - | 7, 8 | 882 | ||
| AY854152.1 | - | 7, 8 | 932 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc7| Gene Symbol: Ccdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029061.3 | NP_083337.1 | 7, 8 | 900 | ||
| GenBank mRNA | AK015811.1 | - | 7, 8 | 882 | ||
| AY854152.1 | - | 7, 8 | 932 | |||