siRNA Details
| Gene Symbol: Snx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127191.2 | NP_001120663.1 | 2 | 335 | ||
| NM_029068.4 | NP_083344.3 | 2 | 335 | |||
| GenBank mRNA | AK015872.1 | - | 2 | 296 | ||
| AK032055.1 | - | 2 | 292 | |||
| AK043975.1 | - | 2 | 305 | |||
| BC019424.1 | - | 1 | 100 | 5052234 | ||
| BC070431.1 | - | 280 | ||||
| BY014829.1 | - | 2 | 329 | |||

Snx16
Genomic Map