siRNA Details
| Gene Symbol: Trim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029077.4 | NP_083353.1 | 1, 2 | 269 | ||
| GenBank mRNA | AB117644.1 | - | 1, 2 | 280 | ||
| AK017887.1 | - | 1, 2 | 243 | |||
| AK076277.1 | - | 251 | ||||
| AK133220.1 | - | 1, 2 | 192 | |||
| AK148837.1 | - | 1, 2 | 233 | |||
| AK172895.1 | - | 1, 2 | 248 | |||
| BC116738.1 | - | 1, 2 | 268 | 40126057 | ||
| BC119130.1 | - | 1, 2 | 268 | 8733879 | ||

Trim14
Genomic Map