siRNA Details
| Gene Symbol: Mdga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081160.1 | NP_001074629.1 | 13, 14 | 2873 | ||
| GenBank mRNA | AK004706.1 | - | 6, 7 | 883 | ||
| DQ788983.1 | - | 3302 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mdga1| Gene Symbol: Mdga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081160.1 | NP_001074629.1 | 13, 14 | 2873 | ||
| GenBank mRNA | AK004706.1 | - | 6, 7 | 883 | ||
| DQ788983.1 | - | 3302 | ||||