siRNA Details
| Gene Symbol: Yipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205157.1 | NP_001192086.1 | 8, 9 | 951 | ||
| NM_001205158.1 | NP_001192087.1 | 8, 9 | 941 | |||
| NM_138303.2 | NP_612176.1 | 7, 8 | 1089 | |||
| GenBank mRNA | AK004959.1 | - | 7, 8 | 875 | ||
| AK033705.1 | - | 8, 9 | 900 | |||
| AK133631.1 | - | 8, 9 | 911 | |||
| BC002282.1 | - | 7, 8 | 1004 | 3497525 | ||
| BC003289.1 | - | 8, 9 | 882 | 3583087 | ||

Yipf2
Genomic Map