siRNA Details
| Gene Symbol: Yipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205157.1 | NP_001192086.1 | 7 | 639 | ||
| NM_001205158.1 | NP_001192087.1 | 7 | 629 | |||
| NM_138303.2 | NP_612176.1 | 6 | 777 | |||
| GenBank mRNA | AK004959.1 | - | 6 | 563 | ||
| AK033705.1 | - | 7 | 588 | |||
| AK133631.1 | - | 7 | 599 | |||
| BC002282.1 | - | 6 | 692 | 3497525 | ||
| BC003289.1 | - | 7 | 570 | 3583087 | ||

Yipf2
Genomic Map