siRNA Details
| Gene Symbol: Hhatl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029095.2 | NP_083371.2 | 2, 3 | 352 | ||
| GenBank mRNA | AK003605.1 | - | 2, 3 | 229 | ||
| AK157924.1 | - | 2, 3 | 150 | |||
| BC024464.1 | - | 2, 3 | 208 | 4975643 | ||
| BY232310.1 | - | 2 | 327 | |||
| BY290483.1 | - | 2, 3 | 353 | |||
| BY703867.1 | - | 2, 3 | 229 | |||

Hhatl
Genomic Map