siRNA Details
Gene Symbol: Lmbr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029098.3 | NP_083374.1 | 1, 2 | 247 | ||
GenBank mRNA | AK003656.1 | - | 1, 2 | 179 | ||
AK030589.1 | - | 1, 2 | 213 | |||
AK049110.1 | - | 1, 2 | 248 | |||
AK143891.1 | - | 1, 2 | 208 | |||
AK154968.1 | - | 1, 2 | 381 | |||
AK170328.1 | - | 1, 2 | 208 | |||
AY052398.1 | - | 1, 2 | 90 | |||
BC023397.1 | - | 1, 2 | 184 | 5043877 |
