siRNA Details
| Gene Symbol: Wipi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178398.4 | NP_848485.1 | 7 | 823 | ||
| GenBank mRNA | AK043857.1 | - | 6 | 1632 | ||
| AK155113.1 | - | 7 | 853 | |||
| AK169051.1 | - | 7 | 797 | |||
| AK172030.1 | - | 7 | 799 | |||
| AK213114.1 | - | 3 | 167 | |||
| BC044894.1 | - | 7 | 848 | 4235237 | ||

Wipi2
Genomic Map