siRNA Details
| Gene Symbol: 4930402F06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080709.2 | NP_001074178.1 | 5 | 475 | ||
| NM_001272040.1 | NP_001258969.1 | 4 | 391 | |||
| GenBank mRNA | AK015047.1 | - | 5 | 476 | ||
| BC115676.1 | - | 4 | 274 | 40104161 | ||
| BC115677.1 | - | 4 | 274 | 40104162 | ||

4930402F06Rik
Genomic Map