siRNA Details
| Gene Symbol: Morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029112.1 | NP_083388.1 | 2, 3 | 351 | ||
| GenBank mRNA | AK015339.1 | - | 2, 3 | 504 | ||
| AK077141.1 | - | 2, 3 | 548 | |||
| BC019987.1 | - | 2, 3 | 351 | 4458628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Morn3| Gene Symbol: Morn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029112.1 | NP_083388.1 | 2, 3 | 351 | ||
| GenBank mRNA | AK015339.1 | - | 2, 3 | 504 | ||
| AK077141.1 | - | 2, 3 | 548 | |||
| BC019987.1 | - | 2, 3 | 351 | 4458628 | ||