siRNA Details
Gene Symbol: Usp47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133758.3 | NP_598519.2 | 7, 8 | 1019 | ||
NM_177249.3 | NP_796223.2 | 8, 9 | 1079 | |||
GenBank mRNA | AK041541.1 | - | 7, 8 | 999 | ||
AK082114.1 | - | 8, 9 | 1043 | |||
AK151051.1 | - | 7, 8 | 1023 | |||
AK219720.1 | - | 3, 4 | 302 | |||
BC108425.1 | - | 8, 9 | 1020 | 30062566 |
