siRNA Details
| Gene Symbol: Usp47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133758.3 | NP_598519.2 | 2 | 314 | ||
| NM_177249.3 | NP_796223.2 | 3 | 374 | |||
| GenBank mRNA | AK041541.1 | - | 2 | 294 | ||
| AK082114.1 | - | 3 | 337 | |||
| AK083763.1 | - | 3 | 590 | |||
| AK151051.1 | - | 2 | 318 | |||
| BC108425.1 | - | 3 | 315 | 30062566 | ||
| BY160489.1 | - | 2 | 314 | |||
| BY743916.1 | - | 2 | 314 | |||

Usp47
Genomic Map