siRNA Details
| Gene Symbol: Rab11fip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033172.3 | NP_001028344.2 | 2 | 975 | ||
| NM_001164367.1 | NP_001157839.1 | 2 | 403 | |||
| GenBank mRNA | AK015534.1 | - | 2 | 404 | ||
| AK154916.1 | - | 2 | 975 | |||
| BC089010.1 | - | 1 | 234 | |||
| BC139380.1 | - | 2 | 738 | 8862402 | ||
| BC139419.1 | - | 2 | 738 | 8862441 | ||

Rab11fip2
Genomic Map