siRNA Details
| Gene Symbol: Fam63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133858.4 | NP_598619.2 | 7 | 1954 | ||
| NM_199475.1 | NP_955769.1 | 7 | 1954 | |||
| GenBank mRNA | AB075603.1 | - | 6 | 965 | ||
| AK036838.1 | - | 7 | 1946 | |||
| AK169673.1 | - | 7 | 1954 | |||
| AK192867.1 | - | 176 | ||||
| AK215644.1 | - | 2 | 221 | |||
| BC039762.1 | - | 6 | 772 | |||
| BC051048.1 | - | 6 | 837 | |||
| BC057901.1 | - | 5 | 483 | |||
| BC093521.1 | - | 3 | 523 | 4007022 | ||
| BC132301.1 | - | 6 | 1199 | 40130578 | ||
| BC144749.1 | - | 6 | 1199 | 9053297 | ||

Fam63a
Genomic Map