siRNA Details
| Gene Symbol: Mei4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175213.3 | NP_780422.1 | 2 | 318 | ||
| GenBank mRNA | AK043929.1 | - | 2 | 319 | ||
| AK082395.1 | - | 2 | 358 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mei4| Gene Symbol: Mei4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175213.3 | NP_780422.1 | 2 | 318 | ||
| GenBank mRNA | AK043929.1 | - | 2 | 319 | ||
| AK082395.1 | - | 2 | 358 | |||