siRNA Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175214.3 | NP_780423.2 | 8 | 2205 | ||
| GenBank mRNA | AK032912.1 | - | 1 | 60 | ||
| BC116646.1 | - | 6 | 1044 | 40086396 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif27| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175214.3 | NP_780423.2 | 8 | 2205 | ||
| GenBank mRNA | AK032912.1 | - | 1 | 60 | ||
| BC116646.1 | - | 6 | 1044 | 40086396 | ||