siRNA Details
Gene Symbol: 4930505A04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100394.1 | NP_001093864.1 | 3 | 418 | ||
GenBank mRNA | AK015698.1 | - | 4 | 486 | ||
BC049719.1 | - | 3 | 397 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930505A04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100394.1 | NP_001093864.1 | 3 | 418 | ||
GenBank mRNA | AK015698.1 | - | 4 | 486 | ||
BC049719.1 | - | 3 | 397 |