siRNA Details
| Gene Symbol: Lysmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191051.1 | NP_001177980.1 | 2, 3 | 708 | ||
| NM_175215.4 | NP_780424.1 | 2, 3 | 371 | |||
| GenBank mRNA | AK033681.1 | - | 2, 3 | 308 | ||
| AK143241.1 | - | 2, 3 | 709 | |||

Lysmd4
Genomic Map