siRNA Details
| Gene Symbol: 4930519F16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029170.1 | NP_083446.1 | 2 | 139 | ||
| GenBank mRNA | AK015842.1 | - | 2 | 148 | ||
| AK039711.1 | - | 2 | 139 | |||
| BC107248.2 | - | 2 | 141 | 40052342 | ||
| BC107249.1 | - | 2 | 137 | 40052344 | ||
| BC114984.1 | - | 2 | 141 | 40052347 | ||
| BC115753.1 | - | 2 | 139 | 40052345 | ||

4930519F16Rik
Genomic Map