siRNA Details
| Gene Symbol: Rprd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081293.1 | NP_001074762.1 | 11 | 3098 | ||
| GenBank mRNA | AK129150.1 | - | 10 | 2645 | ||
| BC067054.1 | - | 11 | 2903 | 30542928 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rprd2| Gene Symbol: Rprd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081293.1 | NP_001074762.1 | 11 | 3098 | ||
| GenBank mRNA | AK129150.1 | - | 10 | 2645 | ||
| BC067054.1 | - | 11 | 2903 | 30542928 | ||