siRNA Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029186.2 | NP_083462.2 | 2 | 306 | ||
| GenBank mRNA | AK031554.1 | - | 2 | 252 | ||
| AK151797.1 | - | 2 | 281 | |||
| AK161305.1 | - | 2 | 262 | |||
| AK170247.1 | - | 253 | ||||
| BC031169.1 | - | 2 | 251 | |||
| BC058746.1 | - | 2 | 252 | 6399588 | ||

Tmem180
Genomic Map