siRNA Details
| Gene Symbol: Rnf121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029211.2 | NP_083487.2 | 4 | 376 | ||
| GenBank mRNA | AK016037.1 | - | 4 | 607 | ||
| AK034806.1 | - | 4 | 1690 | |||
| AK078509.1 | - | 4 | 337 | |||
| AK159032.1 | - | 4 | 513 | |||
| AK159260.1 | - | 4 | 403 | |||
| AK168821.1 | - | 4 | 389 | |||
| AK169089.1 | - | 4 | 389 | |||
| BC022686.1 | - | 4 | 376 | 4241984 | ||

Rnf121
Genomic Map