siRNA Details
| Gene Symbol: Dusp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173745.5 | NP_776106.1 | 2 | 531 | ||
| GenBank mRNA | AK015917.1 | - | 2 | 515 | ||
| AK052827.1 | - | 1 | 33 | |||
| AK079664.1 | - | 2 | 527 | |||
| AK081916.1 | - | 2 | 490 | |||
| BC020036.1 | - | 2 | 525 | 3990751 | ||

Dusp18
Genomic Map