siRNA Details
| Gene Symbol: Pelp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029231.4 | NP_083507.3 | 12 | 1309 | ||
| GenBank mRNA | AK005027.1 | - | 12 | 1307 | ||
| AK128966.1 | - | 12 | 1309 | |||
| BC016444.1 | - | 2 | 126 | 4459875 | ||
| BC057987.1 | - | 6 | 537 | |||
| BC090620.1 | - | 12 | 1303 | 30699968 | ||

Pelp1
Genomic Map