siRNA Details
| Gene Symbol: Prkd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171004.1 | NP_001164475.1 | 19 | 3420 | ||
| NM_001171005.1 | NP_001164476.1 | 18 | 2616 | |||
| NM_029239.3 | NP_083515.2 | 19 | 3417 | |||
| GenBank mRNA | AK030058.1 | - | 18 | 2612 | ||
| BC023383.1 | - | 3 | 488 | |||
| BC027320.1 | - | 18 | 2819 | |||
| BC037012.1 | - | 18 | 3061 | 5342428 | ||
| BC043660.1 | - | 5 | 730 | |||
| BC089164.1 | - | 19 | 3319 | 5400250 | ||

Prkd3
Genomic Map