siRNA Details
| Gene Symbol: Asxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270988.1 | NP_001257917.1 | 12 | 3432 | ||
| NM_172421.5 | NP_766009.2 | 11 | 3288 | |||
| GenBank mRNA | AK036839.1 | - | 11 | 3245 | ||
| AK122540.1 | - | 3 | 2061 | |||
| AK162455.1 | - | 3 | 2029 | |||
| BC141140.1 | - | 12 | 3266 | 9055752 | ||

Asxl2
Genomic Map