siRNA Details
| Gene Symbol: Arhgap36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081123.1 | NP_001074592.1 | 5 | 839 | ||
| GenBank mRNA | AB073967.1 | - | 5 | 706 | ||
| AK048167.1 | - | 5 | 728 | |||
| BC090672.1 | - | 5 | 810 | 30941825 | ||
| BC145645.1 | - | 5 | 805 | 40130948 | ||

Arhgap36
Genomic Map