siRNA Details
| Gene Symbol: Secisbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012732.1 | - | 7 | 973 | ||
| AK030021.1 | - | 7 | 943 | |||
| AK037988.1 | - | 7 | 973 | |||
| AK156076.1 | - | 7 | 1019 | |||
| AK168634.1 | - | 7 | 976 | |||
| BC125391.1 | - | 7 | 999 | 40129906 | ||
| BC125393.1 | - | 7 | 996 | 40129908 | ||
| BC144795.1 | - | 7 | 994 | |||

Secisbp2
Genomic Map