siRNA Details
| Gene Symbol: Secisbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030021.1 | - | 9 | 1258 | ||
| AK156076.1 | - | 9 | 1334 | |||
| AK168634.1 | - | 9 | 1291 | |||
| BC125391.1 | - | 9 | 1314 | 40129906 | ||
| BC125393.1 | - | 9 | 1311 | 40129908 | ||
| BC144795.1 | - | 9 | 1309 | |||

Secisbp2
Genomic Map