siRNA Details
| Gene Symbol: Igfbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018741.2 | NP_061211.1 | 2 | 514 | ||
| GenBank mRNA | AB006141.1 | - | 2 | 502 | ||
| AK012705.1 | - | 2 | 515 | |||
| BC051960.1 | - | 2 | 484 | 5710646 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbpl1| Gene Symbol: Igfbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018741.2 | NP_061211.1 | 2 | 514 | ||
| GenBank mRNA | AB006141.1 | - | 2 | 502 | ||
| AK012705.1 | - | 2 | 515 | |||
| BC051960.1 | - | 2 | 484 | 5710646 | ||