siRNA Details
| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029291.1 | NP_083567.1 | 14, 15 | 1604 | ||
| GenBank mRNA | AK148105.1 | - | 14, 15 | 1636 | ||
| AK207287.1 | - | 260 | ||||
| BC013537.1 | - | 14, 15 | 1604 | 4240784 | ||
| BC024840.1 | - | 7, 8 | 755 | |||

Ascc2
Genomic Map