siRNA Details
| Gene Symbol: 1700001C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029296.2 | NP_083572.1 | 3 | 396 | ||
| GenBank mRNA | BC028263.1 | - | 3 | 359 | 5124927 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001C19Rik| Gene Symbol: 1700001C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029296.2 | NP_083572.1 | 3 | 396 | ||
| GenBank mRNA | BC028263.1 | - | 3 | 359 | 5124927 | |