siRNA Details
| Gene Symbol: Spata19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB120716.1 | - | 2 | 149 | ||
| AK005610.1 | - | 2 | 149 | |||
| JX089334.1 | - | 1 | 321 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata19| Gene Symbol: Spata19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB120716.1 | - | 2 | 149 | ||
| AK005610.1 | - | 2 | 149 | |||
| JX089334.1 | - | 1 | 321 | |||