siRNA Details
Gene Symbol: Ccdc101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029339.3 | NP_083615.3 | 3, 4 | 339 | ||
GenBank mRNA | AK006287.1 | - | 3, 4 | 249 | ||
AK139102.1 | - | 3, 4 | 340 | |||
AK169898.1 | - | 3, 4 | 335 | |||
BC026784.1 | - | 3, 4 | 277 | 4206090 |
