siRNA Details
| Gene Symbol: Ccdc101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029339.3 | NP_083615.3 | 4, 5 | 425 | ||
| GenBank mRNA | AK006287.1 | - | 4, 5 | 335 | ||
| AK139102.1 | - | 4, 5 | 426 | |||
| AK169898.1 | - | 4, 5 | 421 | |||
| BC026784.1 | - | 4, 5 | 363 | 4206090 | ||

Ccdc101
Genomic Map