siRNA Details
| Gene Symbol: Wnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290311.1 | NP_001277240.1 | 21, 22 | 5349 | ||
| NM_001290313.1 | NP_001277242.1 | 21, 22 | 5349 | |||
| NM_029361.4 | NP_083637.2 | 21, 22 | 5349 | |||
| GenBank mRNA | AK044381.1 | - | 2, 3 | 266 | ||
| AK147550.1 | - | 21, 22 | 5036 | |||
| BC046464.1 | - | 14, 15 | 3148 | |||
| BC055795.1 | - | 2, 3 | 244 | |||
| BC060187.1 | - | 10, 11 | 2380 | |||

Wnk2
Genomic Map