siRNA Details
| Gene Symbol: Spaca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029367.1 | NP_083643.1 | 2 | 362 | ||
| GenBank mRNA | AK006357.1 | - | 3 | 510 | ||
| AY572448.1 | - | 2 | 325 | |||
| AY601763.1 | - | 1 | 45 | |||
| BC100503.1 | - | 2 | 362 | 30917230 | ||

Spaca3
Genomic Map