siRNA Details
| Gene Symbol: Snapc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178392.4 | NP_848479.1 | 5 | 700 | ||
| GenBank mRNA | AK012317.1 | - | 4 | 357 | ||
| AK138261.1 | - | 5 | 701 | |||
| AK152833.1 | - | 5 | 706 | |||
| BC030456.1 | - | 5 | 682 | 5401615 | ||

Snapc1
Genomic Map