siRNA Details
| Gene Symbol: Fam65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081241.2 | NP_001074710.2 | 10, 11 | 943 | ||
| GenBank mRNA | AK052316.1 | - | 10, 11 | 892 | ||
| AK122573.1 | - | 10, 11 | 917 | |||
| BC079880.1 | - | 10, 11 | 857 | 6852251 | ||

Fam65a
Genomic Map