siRNA Details
| Gene Symbol: Morc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193309.1 | NP_001180238.1 | 16 | 2810 | ||
| NM_029413.4 | NP_083689.2 | 16 | 2810 | |||
| GenBank mRNA | AK032807.1 | - | 16 | 2811 | ||
| AK160137.1 | - | 2 | 249 | |||
| BC036164.1 | - | 10 | 1696 | |||
| BC039283.1 | - | 9 | 1522 | |||
| BC098483.1 | - | 16 | 2765 | 30022418 | ||
| BC132153.1 | - | 16 | 2664 | 40130430 | ||
| BC132497.1 | - | 16 | 2664 | 40130774 | ||

Morc4
Genomic Map