siRNA Details
| Gene Symbol: Dcstamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289506.1 | NP_001276435.1 | 2 | 562 | ||
| NM_001289508.1 | NP_001276437.1 | 2 | 562 | |||
| NM_001289512.1 | NP_001276441.1 | 2 | 562 | |||
| NM_001289513.1 | NP_001276442.1 | 2 | 562 | |||
| NM_029422.4 | NP_083698.1 | 2 | 562 | |||
| GenBank mRNA | AB109560.1 | - | 2 | 560 | ||
| AB109561.1 | - | 2 | 560 | |||
| AK014697.1 | - | 2 | 561 | |||
| AK154384.1 | - | 2 | 561 | |||
| AY517483.1 | - | 2 | 561 | |||
| AY517485.1 | - | 2 | 561 | |||
| BC139146.1 | - | 2 | 548 | 8862168 | ||
| BC145557.1 | - | 2 | 548 | |||
| BC145558.1 | - | 2 | 548 | 9054122 | ||

Dcstamp
Genomic Map