siRNA Details
| Gene Symbol: Rab11fip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080813.2 | NP_001074282.1 | 3 | 1015 | ||
| NM_029423.2 | NP_083699.2 | 3 | 1015 | |||
| GenBank mRNA | AK014696.1 | - | 3 | 997 | ||
| AK030769.1 | - | 3 | 1018 | |||
| AK086797.1 | - | 3 | 1013 | |||
| AK151024.1 | - | 3 | 1016 | |||
| BC091001.1 | - | 2 | 454 | |||
| BC125400.1 | - | 3 | 1000 | 40129915 | ||
| BC132094.1 | - | 3 | 1000 | 40130371 | ||

Rab11fip1
Genomic Map