siRNA Details
Gene Symbol: Lppr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014763.1 | - | 3 | 520 | ||
AK032034.1 | - | 3 | 886 | |||
AK043629.1 | - | 3 | 510 | |||
AK076176.1 | - | 3 | 651 | |||
AK082692.1 | - | 3 | 782 | |||
AY512657.1 | - | 3 | 433 | |||
BC031879.1 | - | 3 | 750 | 4527789 |
