siRNA Details
| Gene Symbol: Klhl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029436.3 | NP_083712.4 | 3 | 344 | ||
| GenBank mRNA | AK015239.1 | - | 3 | 330 | ||
| AK044892.1 | - | 3 | 316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl24| Gene Symbol: Klhl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029436.3 | NP_083712.4 | 3 | 344 | ||
| GenBank mRNA | AK015239.1 | - | 3 | 330 | ||
| AK044892.1 | - | 3 | 316 | |||